Learn More
Corning™ LSE™ Shaking Incubator Accessory, Plastic Clamps
Shop All Corning Products
Click to view available options
Holds:
125 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Description
Constructed of polypropylene.
Specifications
Specifications
| Holds | 250 mL |
| Product Type | Clamp |
| For Use With (Equipment) | LSE Benchtop Shaking Incubator |