Learn More
Novus Biologicals™ LPS from E. Coli, TLR4 ligand
Product Code. p-200087446 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Product Code. 18656528
Brand: Novus Biologicals™ NBP2252951.0mg
Applications: Functional
Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs). Use in functional reported in scientific literature (PMID 26121241).
Lipopolysaccharides from Escherichia coli 0111:B4 Gamma-irradiated, BioXtra, suitable for cell culture Dilution: Functional Purity: for ion exchange chromatography Reconstitute with 1 ml sterile balanced salt solution or tissue culture medium to the vial (1 mg) and gently swirl until the powder dissolves. Reconstituted product may be further diluted to desired working concentrations.
TRUSTED_SUSTAINABILITY
Specifications
Concentration LYOPH
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Quantity 1 mg
For Use With (Application) Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs). Use in functional reported in scientific literature (PMID 26121241).
Target LPS from E. Coli, TLR4 Ligand