Learn More
Buerkle™ Lowering Cable for Buerkle™ Dipping Bottles
Product Code. 10579902 Shop All Bürkle Products
Click to view available options
Length (Metric):
10 m
100 m
25 m
50 m
Material:
Copper
Polypropylene
Stainless Steel
Stainless Steel (V2A)
Diameter (Metric) Outer:
1.25 mm
2 mm
2.3 mm
4.5 mm
Unit Size:
Each
Product Code. 10579902
Brand: Buerkle™ 53090510
Buerkle™ Lowering Cable for dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials.
  • Good chemical resistance
  • For dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials
  • Tensile load max. up to 10kg
TRUSTED_SUSTAINABILITY
Specifications
Product Type Lowering Cable
Material Stainless Steel
For Use With (Application) Dipping Bottles and Immersion Cylinders, Manually Operated Drum Reels
Diameter (Metric) Outer 2.3 mm
Length (Metric) 10 m