Learn More
Buerkle™ Lowering Cable for Buerkle™ Dipping Bottles
Shop All Bürkle Products
Click to view available options
Length (Metric):
10 m
100 m
25 m
50 m
Material:
Copper
Polypropylene
Stainless Steel
Stainless Steel (V2A)
Diameter (Metric) Outer:
1.25 mm
2 mm
2.3 mm
4.5 mm
Unit Size:
Each
Description
- Good chemical resistance
- For dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials
- Tensile load max. up to 10kg
Specifications
Specifications
| Product Type | Lowering Cable |
| Material | Stainless Steel (V2A) |
| For Use With (Application) | Dipping Samplers and Immersion Cylinders, Manually-Operated Drum Reels |
| Diameter (Metric) Outer | 1.25 mm |
| Length (Metric) | 10 m |