Learn More
Scotch-Brite™ Low Scratch Scouring Pad, 2000HEX
Product Code. 17118860
Change view
Click to view available options
Dimensions (L x W):
146 x 127 mm
Unit Size:
Pack of 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17118860 146 x 127 mm Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17118860 Supplier Scotch-Brite™ Supplier No. 7100197274
  • ONLINE EXCLUSIVE PRICE
    28.00€
    15.44€ / Pack of 15
    Save 12.56€
    44% Off
View all promotions
Estimated Shipment: 20-11-2026
A powerful, 2 in 1 scouring pad that features a raised, medium duty power dot side. Engineered to scour away tough messes like a medium duty pad, but with less visible scratches.
  • Designed to clean a wide variety of surfaces where minimal scratching is desired
  • The dark blue side removes fine food soil, leaving behind a virtually spotless finish
  • Ergonomic hexagon shape fits comfortably in the hand
  • Easily cleans hard to reach areas
  • HACCP certified
TRUSTED_SUSTAINABILITY
Specifications
Product Type Scouring Pad
Dimensions (L x W) 146 x 127 mm
For Use With (Application) Commercial Kitchen Cleaning
Length (Metric) 146 mm
Model 2000 HEX
Quantity 15 Pk.
Style Low Scratch
Width (Metric) 127 mm