Learn More
Thermo Scientific™ Low Profile Strip Tubes without Caps Promotion
Product Code. 15717486
Click to view available options
Color:
Natural
Unit Size:
Pack of 120
Product Code. 15717486
Brand: Thermo Scientific™ AB0771
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Increase the efficiency of reactions with these low-profile strip tubes for PCR and qPCR applications
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Certified free of human DNA, RNase, DNase, and endotoxins
Color Natural
Description PCR, qPCR and Real-Time PCR applications
Quantity Strips of 8; 10 Packs of 12