Learn More
Porcelaines Avignon™ Low Form Porcelain Crucibles
Click to view available options
Capacity (Metric):
3 mL
6 mL
14 mL
25 mL
40 mL
80 mL
130 mL
250 mL
Unit Size:
Pack of 6
Description
- Low walls
- Withstand high temperatures and rapid temperature changes
- Porcelain
Specifications
Specifications
| Material | Porcelain |
| Finish | Glazed |
| Capacity (Metric) | 6 mL |
| Diameter (Metric) Outer | 33mm |
| Height (Metric) | 21mm |
| Product Type | Crucible |