Learn More
Porcelaines Avignon™ Low Form Porcelain Crucibles
Product Code. 11758463
Click to view available options
Capacity (Metric):
3 mL
6 mL
14 mL
25 mL
40 mL
80 mL
130 mL
250 mL
Unit Size:
Pack of 6
Product Code. 11758463
Brand: Porcelaines Avignon™ 11301
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Heat and cool substances in the laboratory using Porcelaines Avignon™ Low Form Porcelain Crucibles. They are available in various capacities.
  • Low walls
  • Withstand high temperatures and rapid temperature changes
  • Porcelain
Specifications
Material Porcelain
Capacity (Metric) 3 mL
Diameter (Metric) Outer 25mm
Height (Metric) 18mm
Product Type Crucible