Learn More
Witeg™ Low Form Glass Beakers with Spout
Click to view available options
Capacity (Metric):
150 mL
250 mL
400 mL
600 mL
5000 mL
Unit Size:
Pack of 10
Description
- Wide range of capacities available
- Acc. to ISO 3819
- Manufactured from DURAN™ borosilicate glass 3.3
- With spout for easy pouring
Specifications
Specifications
| Color | Transparent |
| Diameter (Metric) Outer | 80 mm |
| Capacity (Metric) | 400 mL |
| Material | Borosilicate Glass |
| Height (Metric) | 110 mm |
| Includes | Spout |
| Type | Beaker |
| Graduated | Yes |
| Certifications/Compliance | ISO 3819 |
| Style | Low Form |