missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Low Fluorescence Glass Plate Kit
Shop All Cytiva Products
Click to view available options
Unit Size:
1 set
Description
Includes
- One borosilicate glass plate (27.6 x 21.7cm x 3mm)
- One borosilicate glass plate (27.6 x 21.1cm x 3mm)
- Two vinyl spacer (10mm x 1mm)
Specifications
Specifications
| Type | Low Fluorescence Glass Plate Kit |
| Includes | 1 Borosilicate Glass plate (27.6 x 21.7cm x 3mm), 1 Borosilicate Glass plate (27.6 x 21.1cm x 3mm), 2 Vinyl spacer (10 x 1mm) |
| Quantity | Complete packsize |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction