Learn More
Lovibond™ Total Nitrogen Reagent Set, for Tube Testing, Low Range
Product Code. 15268093
Change view
Click to view available options
Range:
0 to 25mg/L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
15268093 0 to 25mg/L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15268093 Supplier Lovibond™ Supplier No. 535550
Includes
Total N Persulfate Reagent, Total Nitrogen Reagent A, Total Nitrogen Reagent B, Total Nitrogen Hydroxide LR Tubes, Total Nitrogen Acid LR/HR Tubes, Deionized Water, 100mL
Premeasured unit dose reagent for water quality testing
  • Offers same high quality standards as tablet reagents
  • Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open package
  • Easily create “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) Hach™ and other manufacturers test kits, colorimeters and spectrophotometers, reactor
Includes Total N persulfate reagent (50 pack), total nitrogen reagent A (50 pack), total nitrogen reagent B (50 pack), 50 ct. total nitrogen hydroxide LR tubes, 50 ct. total nitrogen acid LR/HR tubes, 100mL deionized water
No. of Tests 25
Range 0 to 25mg/L
Test Method Persulfate Digestion
Type Low Range Total Nitrogen Reagent Set
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • vario total nitrogen LR set
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • Oxidising solid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: disodium disulphite,disodium 4,5-dihydroxynapthalene-2,7-disulphonate, quartz