Learn More
Lovibond™ ET 740 and ET 750 - Laboratory Floc / Jar Tester
Product Code. 12651745
Change view
Click to view available options
Length (Metric):
64.7 cm
93.9 cm
Weight (Metric):
12.1 kg
17 kg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Weight (Metric) unitSize
12651745 64.7 cm 12.1 kg Each
12661745 93.9 cm 17 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12651745 Supplier Lovibond™ Supplier No. 2419155
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Includes
Power Supply with US, EU and UK adapter. Instruction Manual.
Requires
1,000 mL beakers, Low form
Flocculation Testers (also known as Jar Testers) are designed for a range of applications - such as testing the efficiency of flocculation or precipitation agents
Continuously variable stirring speed (lab versions only.)
  • Digital display of stirring RPM.
  • Timer function.
  • Illuminated back panel.
  • Height adjustment option for the stirring blades during operation.
  • Date and Time.
Floc Testers are not supplied with beakers. Please purchase from a laboratory products supplier.
Specifications
Type Laboratory Flocculation Tester
Display Type Digital
Includes Power supply with US, EU and UK adapter, instruction manual
Key Functions Timer
For Use With (Application) For a range of applications such as testing the efficiency of flocculation or precipitation agents
Resolution 1 Revolution
Dimensions (L x W x H) 25.5 x 13.5 x 10.25 in. (64.7 x 34.2 x 26.03 cm)
Length (English) 25.5 in.
Length (Metric) 64.7 cm
Width (English) 13.5 in.
Width (Metric) 34.2 cm
Height (English) 10.25 in.
Height (Metric) 26.03 cm
Electrical Requirements 115/230 V 50/60 Hz
Frequency 50/60 Hz
Voltage 115/230 V
Weight (English) 26.8 lb.
Weight (Metric) 12.1 kg