missing translation for 'onlineSavingsMsg'
Learn More

Loperamide Hydrochloride, TRC

Product Code. 18022720
Change view
Click to view available options
Quantity:
5 mg
50 mg
1 g
Unit Size:
1g
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18022720 1 g 1g
18002220 50 mg 50mg
18041220 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18022720 Supplier Toronto Research Chemicals Supplier No. TRCL4694501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34552-83-5
Molecular Formula C29 H33 Cl N2 O2 . Cl H
Molecular Weight (g/mol) 513.5
Synonym 4-(4-Chlorophenyl)-4-hydroxy-N,N-dimethyl-alpha,alpha-diphenyl-1-piperidinebutanamide monohydrochloride, 4-(p-Chlorophenyl)-4-hydroxy-N,N-dimethyl-alpha,alpha-diphenyl-1-piperidinobutyramide hydrochloride, Arret, Blox, Brek, Dissenten, Fortasec, Imodium, Imodium A-D, Imosec, Imossel, Lopemid, Lopemin, Loperamide hydrochloride, Loperazine, Loperyl, Lopodium, Maalox Antidiarrheal, NSC 696356, PJ 185, R 18553, Suprasec, Tebloc
IUPAC Name 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride
SMILES Cl.CN(C)C(=O)C(CCN1CCC(O)(CC1)c2ccc(Cl)cc2)(c3ccccc3)c4ccccc4

Specifications

Quantity 1 g
InChI Formula InChI=1S/C29H33ClN2O2.ClH/c1-31(2)27(33)29(24-9-5-3-6-10-24,25-11-7-4-8-12-25)19-22-32-20-17-28(34,18-21-32)23-13-15-26(30)16-14-23;/h3-16,34H,17-22H2,1-2H3;1H
Formula Weight 512.1997
Percent Purity >95%
Chemical Name or Material Loperamide Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.