Learn More
Kappes Systeme™ Longitudinal Divider
Click to view available options
Height (Metric):
105 mm
145 mm
Unit Size:
Each
Specifications
Specifications
| Product Type | Longitudinal Divider |
| Color | Zinc |
| For Use With (Equipment) | Secure and professional storage |
| Quantity | 1/Pk. |
| Height (Metric) | 105 mm |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido