Learn More
Hamilton™ Long Life Instrument Syringes
Long life syringes were developed for extreme conditions that include harsh chemicals as well as abrasive particulate matter.
188.00€ - 438.00€
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11580232
|
Hamilton™
8300-25 |
250 μL |
188.00€
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
10422385
|
Hamilton™
8300-45 |
5 mL |
284.00€
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
10034194
|
Hamilton™
8250-20 |
50 μL |
212.00€
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
10094234
|
Hamilton™
8250-25 |
150 μL |
236.00€
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
10176164
|
Hamilton™
8250-15 |
25 μL |
438.00€
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
10420465
|
Hamilton™
8250-35 |
500 μL |
228.00€
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
10698025
|
Hamilton™
8250-22 |
100 μL |
233.00€
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
10748965
|
Hamilton™
8300-35 |
1 mL |
195.00€
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Description
- Plunger tip made from UHMWPE (Ultra High Molecular Weight Poly Ethylene) which is harder than PTFE to better resist scratching
- Plunger rod made from 316 stainless steel which offers exceptional chemical resistance
Specifications
1/Ea. | |
Luer Lock |