Description
Clean sensitive surfaces with ease, thanks to their extreme low-lint properties.
Reduce waste, as these strong cleaning cloths won't tear easily and can be reused.
Tork Long Lasting Cleaning Cloths White W8:
Save on space and costs with these long-lasting cleaning cloths. They're strong enough to be reused and won't tear easily. Plus, their multipurpose and highly absorbent design means they can tackle a wide range of tasks with ease, including stubborn spills and marks. Low-lint properties even make the cleaning cloths practical for sensitive surfaces. Use with the Tork Folded Wiper/Cloth Dispenser to protect the refills. Its one-at-a-time dispensing feature results in less consumption and waste.
Advantages at a glance:
- Compatible with most solvents, disinfectants and detergents
- Suitable for a wide range of tasks
- Extremely low-lint for sensitive surfaces
- Strong and absorbent for easy reuse
Specifications
Specifications
| Product Type | Cleaning Cloth |
| Length (English) | 15.16 in. |
| Length (Metric) | 38.5 cm |
| Width (English) | 11.81 in. |
| Width (Metric) | 30 cm |
| Material | Paper |
| Certifications/Compliance | ISO 9001, ISO 14001, FSC MIX Credit |
| Color | White |
| Disposable | Reusable |
| Packaging Quantity | 400/Cs. |
| Show More |