Learn More
Lenz Laborglasintrumente Long Ground Cones With Tip
Product Code. p-7222503
Click to view available options
Joint Size:
NS 14/35
NS 19/38
NS 24/40
NS 29/42
Unit Size:
Pack of 10
Product Code. 10090291
Brand: Lenz Laborglasintrumente 01100824
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Conical ground cones
  • Made of DURAN™ tubing
Specifications
Quantity 10/Pk.
Joint Size NS 24/40
Thickness (Metric) Wall 1.8 mm
Diameter (Metric) Outer Tube 22 mm
Length (Metric) 130 mm
Material Borosilicate Glass (3.3)