missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHOWA™ Long Cuffed Nitrile Gloves, Model 7565
Description
- By chlorinating the glove, comfort is enhanced and the feeling of tackiness is removed.
- The nitrile construction protects against chemical penetration and projection.
- Free of latex, powder and silicone to prevent allergic reaction and skin irritation.
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 0 |
| Chlorinated | Yes |
| Color | Black |
| Finger Thickness (English) | 5.9 mil |
| Finger Thickness (Metric) | 0.15 mm |
| Latex Free | Yes |
| Material | Biodegradable Nitrile |
| Palm Thickness (English) | 5.9 mil |
| Palm Thickness (Metric) | 0.15 mm |
| Size | 2X-Large |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?