Learn More
Brady™ Lockout Duffel Bag
Product Code. 11761955 Shop All Brady Products
Change view
Click to view available options
Color:
Blue, Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11761955 Blue, Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11761955 Supplier Brady™ Supplier No. 099162
  • ONLINE EXCLUSIVE PRICE
    112.00€
    108.66€ / Pack of 1
    Save 3.34€
    2% Off
In Stock
Create your own lockout kits by filling the duffel bag with the contents of your choice. The Lockout Duffel Bag is made of 600 Denier Polyester with 95 cm removable shoulder strap and 8 inside pockets.
  • Contents not included.
Specifications
Product Type Duffel Bag
Color Blue, Black
Material 600D Polyester
Depth (English) 8.5 in.
Depth (Metric) 215.90 mm
Description Lockout Duffel Bag
Dimensions (L x W) 420 x 215 mm
For Use With (Application) Lockout Tagout
Height (English) 10 in.
Height (Metric) 254 mm
Quantity 1 Ea.
Width (English) 16.5 in.
Width (Metric) 419.1 mm