Learn More
Lobeline sulfate, 99%, ACROS Organics™
Product Code. 10632223 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10632223 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10632223 Supplier Thermo Scientific Acros Supplier No. 352640010
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
CAS 134-64-5
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C44H56N2O8S
MDL Number MFCD00135592
Quantity 1 g
Merck Index 15, 5609
Specific Rotation Condition −25° (20°C c=2)
Synonym lobeline sulfate, bantron, lobelin sulfate, lobeline sulphate, unii-4cj480v2hp, lobeline, sulfate 2:1 salt, ethanone, 2-6-2-hydroxy-2-phenylethyl-1-methyl-2-piperidinyl-1-phenyl-, 2theta-2alpha,6alpha s-, sulfate 2:1 salt, bis--lobeline ; sulfuric acid, smokeless tn, 2-6-2-hydroxy-2-phenylethyl-1-methylpiperidin-2-yl-1-phenylethanone sulfate
InChI Key GRZMOSSVIPFGFF-GNJLJDPWSA-N
SMILES OS(O)(=O)=O.CN1[C@H](C[C@H](O)C2=CC=CC=C2)CCC[C@@H]1CC(=O)C1=CC=CC=C1.CN1[C@H](C[C@H](O)C2=CC=CC=C2)CCC[C@@H]1CC(=O)C1=CC=CC=C1
IUPAC Name bis(2-[(2R,6S)-6-[(2S)-2-hydroxy-2-phenylethyl]-1-methylpiperidin-2-yl]-1-phenylethan-1-one); sulfuric acid
Molecular Weight (g/mol) 773.00
PubChem CID 120636
Formula Weight 386.51
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material Lobeline sulfate, 99%