Learn More
Brannan™ LO-tox™ Lab Thermometer
Product Code. 13038583
Change view
Click to view available options
Temperature Range (Metric):
-1°C to +101°C
-1°C to +51°C
-10°C to +101°C
-10°C to +110°C
-10°C to +210°C
-10°C to +260°C
-10°C to +300°C
-10°C to +330°C
-10°C to +360°C
-10°C to +50°C
-20°C to +110°C
-20°C to +150°C
-35°C to +50°C
Subdivision:
0.1°C
0.2°C
0.5°C
1°C
2°C
Immersion Type:
Partial Immersion
Total Immersion
Length (Metric):
115 mm
155 mm
305 mm
405 mm
Unit Size:
Each
Pack of 10
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Subdivision Immersion Type Length (Metric) Unit Size
13038583 -10°C to +210°C 1°C Partial Immersion 405 mm Each
13098583 -35°C to +50°C 1°C Partial Immersion 305 mm Each
13018593 -20°C to +110°C 1°C Partial Immersion 305 mm Each
13028593 -20°C to +150°C 1°C Partial Immersion 305 mm Each
11907341 -10°C to +260°C 1°C Partial Immersion 405 mm Each
12309359 -10°C to +50°C 0.5°C Total Immersion 305 mm Each
12399349 -35°C to +50°C 1°C Total Immersion 305 mm Each
13078583 -20°C to +110°C 1°C Total Immersion 305 mm Each
13088583 -20°C to +150°C 1°C Total Immersion 305 mm Each
12389349 -10°C to +210°C 1°C Total Immersion 405 mm Each
13068593 -10°C to +330°C 2°C Partial Immersion 305 mm Each
12319359 -10°C to +300°C 1°C Partial Immersion 305 mm Each
13038603 -1°C to +51°C 0.1°C Partial Immersion 405 mm Each
13008603 -1°C to +51°C 0.1°C Total Immersion 405 mm Each
13048603 -1°C to +51°C 0.2°C Partial Immersion 405 mm Each
13018603 -1°C to +51°C 0.2°C Total Immersion 405 mm Each
13058603 -10°C to +101°C 0.2°C Partial Immersion 405 mm Each
12339359 -1°C to +101°C 0.2°C Total Immersion 405 mm Each
13038593 -10°C to +110°C 1°C Total Immersion 155 mm Each
13048593 -10°C to +50°C 1°C Total Immersion 115 mm Each
15482695 -10°C to +360°C 2°C Partial Immersion 305 mm Pack of 10
15442695 -20°C to +110°C 0.5°C Total Immersion 305 mm Pack of 10
15432695 -10°C to +360°C 2°C Total Immersion 305 mm Pack of 10
15689025 -20°C to +110°C 0.5°C Partial Immersion 305 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Product Code. 13038583 Supplier Brannan™ Supplier No. 44/805/8
  • ONLINE EXCLUSIVE PRICE
    4.35€
    3.97€ / Pack of 1
    Save 0.38€
    8% Off
View all promotions
 Restricted Item
Estimated Shipment: 04-12-2026
LO-tox™ lab thermometer without calibration certificate.
Read temperatures quickly and accurately with blue spirit against a white background. Brannan™ Lo-Tox™ Spirit-filled partial Immersion Thermometer is manufactured to traceable standards and includes a blue non-roll top and safety suspension ring.
  • General purpose/stirring thermometer with a low toxic, environmentally friendly filling
  • Easy to read against background
  • Each thermometer is manufactured to traceable standards
Specifications
Bottle Fill Blue Spirit
For Use With (Application) General use
Immersion Type Partial Immersion
Includes Brannan non roll end cap
Length (Metric) 405 mm
Material Glass
Product Type Laboratory Thermometer
Max. Temperature (Metric) 210°C
Min. Temperature (Metric) -10°C
Temperature Range (Metric) -10°C to +210°C
Accuracy ±2°C
Subdivision 1°C
Immersion Length 76 mm