Learn More
Tocris Bioscience™ LNT 1
Description
LNT 1 is a potent flap endonuclease 1 (FEN1) inhibitor (IC50 = 46.4 nM for hFEN1-336Δ). Cytotoxic to SW620 colorectal cancer cells in vitro; induces DNA damage response.
Specifications
Specifications
| Chemical Name or Material | 1-[(2,3-Dihydro-1,4-benzodioxin-2-yl)methyl]-3-hydroxythieno[3,2-d]pyrimidine-2,4(1H,3H)-dione |
| CAS | 824983-91-7 |
| Quantity | 5 mg |
| Molecular Formula | C15H12N2O5S |
| Formula Weight | 332.33 |
| Purity | 98% |