Learn More
Thermo Scientific™ LN2 Level Monitor
Product Code. p-4530542 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Locator 4 Plus
Locator 8 Plus
Locator Jr. Plus
Voltage:
100/240 V
230 V
Unit Size:
Each
Product Code. 10174623
Brand: Thermo Scientific™ CN509X170
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Safeguard irreplaceable biological specimens with the LN2 Level Monitor, one of the most advanced liquid nitrogen level monitors available.
Safeguard irreplaceable biological specimens with the Thermo Scientific™ LN2 Level Monitor, one of the most advanced liquid nitrogen level monitors available. Designed to minimize liquid nitrogen evaporation, the monitor is for use with Thermo Scientific™ Locator™ and Locator Plus cryogenic storage systems.
Ultrasonic Level Monitor
  • Minimizes conduction and liquid nitrogen evaporation
  • Easy-to-read indicator continuously displays liquid nitrogen volume in 1/8 increments
  • Audible and visual alarms signal low level liquid nitrogen conditions
TRUSTED_SUSTAINABILITY
Specifications
Description Level Monitor
Type LN2 Measuring Device
For Use With (Equipment) Locator Jr. Plus
Monitoring Options Alarm
Voltage 100/240 V