Learn More
Abnova™ LMO7 Split FISH Probe
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark |
| Format | Tube |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
| Gene | LIM domain 7 |
| Gene Alias | FBX20,FBXO20,KIAA0858,LOMP |
| Gene Symbol | LMO7 |
| Gene ID (Entrez) | 4008 |
| Product Type | DNA/RNA |
| Quantity | 100 μL |
| Form | Liquid |