Learn More
LMBRD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554021
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KGTRSAAYER LENTEDIEEV EQHIQTIKSK SKDGRPLPAR DKRALKQFEE RLRTLKKRER HLEFIENSWW TKFCGALRP Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 91%.
LMBRD1, also known as NESI (nuclear export signal-interacting protein, is a lysosomal membrane protein that is thought be involved in the transport and metabolism of cobalamin. LMBRD1 was initially identified as interacting with the large form of the hepatitis delta antigen and may be required for the nucleocytoplasmic shuttling of the hepatitis delta virus. Mutations in this gene are associated with the vitamin B12 metabolism disorder termed, homocystinuria-megaloblastic anemia complementation type F (cblF).Specifications
LMBRD1 | |
Polyclonal | |
Unconjugated | |
Lmbrd1 | |
0910001K20Rik; AV347960; BM-021; C6orf209; CD001; HDAg-L-interacting protein NESI; hepatitis delta antigen-L interacting protein; liver regeneration p-53 related protein; LMBD1; LMBR1 domain containing 1; LMBR1 domain-containing protein 1; LMBRD1; MAHCF; MSTP044; NESI; nuclear export signal-interacting protein; Probable lysosomal cobalamin transporter; protein N90b | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55788 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NUN5 | |
Lmbrd1 | |
Recombinant protein corresponding to Human LMBRD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only