missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LJI308

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
LJI308 is a potent pan-ribosomal S6 protein kinase (RSK) inhibitor (IC50 values are 4 to 13 nM). Displays selectivity for RSK isoforms over other kinases. Inhibits RSK-dependent Y-box binding protein 1 (YB-1) phosphorylation in vitro. Suppresses growth of triple negative breast cancer cell lines.
Specifications
Specifications
| Chemical Name or Material | 2,6-Difluoro-4-[4-[4-(4-morpholinyl)phenyl]-3-pyridinyl]phenol |
| CAS | 1627709-94-7 |
| Quantity | 5 mg |
| Molecular Formula | C21H18F2N2O2 |
| Formula Weight | 368.38 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction