Learn More
Lithium tetraborate, Puratronic™, 99.998% (metals basis)
Description
Used in the laboratory as LB (Lithium borate) buffer for gel electrophoresis of DNA and RNA. Employed in the borax fusion method to modify mineral powder samples for analysis by WDXRF (Wavelength Dispersive X-Ray Fluorescence) Spectroscopy. Acts as an ingredient in the manufacture of glasses and ceramics. Useful fod dissolving basic oxides such as CaO and MgO. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 930°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 1 kg |
| Assay Percent Range | 99.998% (Metals Basis) |
| Formula Weight | 169.12 |
| Density | 2.4g/cm3 at 20°C |
| Chemical Name or Material | Lithium Tetraborate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.