missing translation for 'onlineSavingsMsg'
Learn More

Lithium isopropoxide, 99.9% (metals basis)

Product Code. 11340198
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11340198

Brand: Thermo Scientific Alfa Aesar 042930.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2388-10-5 | C3H7LiO | 66.03 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2388-10-5
Molecular Formula C3H7LiO
Molecular Weight (g/mol) 66.03
MDL Number MFCD00048286
InChI Key HAUKUGBTJXWQMF-UHFFFAOYSA-N
Synonym lithium isopropoxide, lithium propan-2-olate, lithium isopropanolate, lithium 2-propanolate, acmc-1ch0u, lithium isopropoxide solution, 2-propanol, lithiumsalt 1:1, lithium 1+ propan-2-olate
PubChem CID 23673350
IUPAC Name lithium;propan-2-olate
SMILES [Li+].CC(C)[O-]

Specifications

Color White to Yellow
Linear Formula (CH3)2CHOLi
Quantity 10 g
UN Number UN3206
Beilstein 3618614
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 66.03
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Lithium isopropoxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.