missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide monohydrate, Honeywell Fluka™

Product Code. p-7072200 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
500 g
2 kg
Unit Size:
25g
2kg
500g
This item is not returnable. View return policy

Product Code. 15605420

Brand: Honeywell-Fluka 40297425G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
PubChem CID 168937
IUPAC Name lithium;hydroxide;hydrate
SMILES [Li+].O.[OH-]

Specifications

Quantity 25 g
Linear Formula LiOH · H2O
UN Number UN2680
Formula Weight 41.96g/mol
Percent Purity ≥98%
Chemical Name or Material Lithium hydroxide monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.