missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lithium dodecylsulfate, 98%

Product Code. 11418616
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11418616

Brand: Thermo Scientific Chemicals A16005.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2044-56-6 | C12H25LiO4S | 272.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2044-56-6
Molecular Formula C12H25LiO4S
Molecular Weight (g/mol) 272.33
MDL Number MFCD00007467
InChI Key YFVGRULMIQXYNE-UHFFFAOYSA-M
Synonym lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure
PubChem CID 2735071
SMILES [Li+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Quantity 100 g
UN Number UN1325
IUPAC Name lithium;dodecyl sulfate
Formula Weight 272.33
Percent Purity 98%
Odor Odorless
Chemical Name or Material Lithium dodecylsulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.