missing translation for 'onlineSavingsMsg'
Learn More

Lithium bromide, ultra dry, 99.998% (metals basis)

Product Code. 11322488
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11322488

Brand: Thermo Scientific Alfa Aesar 014537.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7550-35-8 | BrLi | 86.844 g/mol

Lithium bromide is used for reconstitution of brines, as swelling agent for proteins and as electrolyte component in lithium battery. Used as brazing and welding flux. Lithium bromide is used as catalyst for the condensation of carbonyl compounds with active methylene compounds in the absence of solvent, operating medium for air-conditioning and industrial drying system. Also used in manufacturing pharmaceuticals and alkylation process. It also plays an important role in the synthesis of dihydropyrimidinones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7550-35-8
Molecular Formula BrLi
Molecular Weight (g/mol) 86.844
MDL Number MFCD00011077
InChI Key AMXOYNBUYSYVKV-UHFFFAOYSA-M
Synonym lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
PubChem CID 82050
ChEBI CHEBI:63042
IUPAC Name lithium;bromide
SMILES [Li+].[Br-]

Specifications

Boiling Point 1265°C
Melting Point 550°C
Color White
Physical Form Beads
Quantity 25 g
Assay Percent Range 99.998% (Metals basis)
Solubility Information Very soluble in water and alcohol. Soluble in methanol,acetone,and glycol. Slightly soluble in pyridine.
Formula Weight 86.85
Sensitivity Hygroscopic
Density 3.464 g/cm3 at 20°C
Chemical Name or Material Lithium bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.