Learn More
Agilent BioTek Liquid Level Alert Kit
Liquid Level Alert System for 50 TS Configurations
Brand:  Agilent BioTek 4070035
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12504906
  • 1075.00€ / Each
Estimated Shipment: 19-08-2024
Description
  • Liquid Level Alert System for 50 TS Configurations without Buffer Switching
  • For non-“V” configurations. Includes select box with one 2L supply bottle and one 2L waste bottle
Specifications
Specifications
Liquid Level Alert Kit
1 Pk.
Select box with three 2-Liter supply bottles and one 2-Liter waste bottle
ELx 50 Models, Non-Buffer Switching