Learn More
AHLSTROM Lipid Saver Card
Description
Lipid Saver™ features five printed 12 mm circles, each capable of holding up to 70 μl of blood, allowing for a maximum collection of 350 μl. Compatible with both manual and semi-automated punching systems, Lipid Saver™ seamlessly integrates with current dried blood spot (DBS) technologies. The collection paper is chemically treated with a proprietary stabilizing solution, preventing oxidative degradation of the analytes and ensuring sample integrity over extended periods.
Key Areas Of Use Include:
- Lipidomics
- Well-being
- Diseases Prevention
- Therapeutic drug monitoring
Specifications
Specifications
| Product Type | Specimen Collection and Transport Card |
| Closure Included | Yes |
| Graduated | No |
| Material | Pure Absorbent Fibers |
| Shape | Rectangular |
| Sterility | Non-sterile |
| Capacity (Metric) | 70 μL/Sample Area |
| Certifications/Compliance | ISO 9001 |