Learn More
AHLSTROM Lipid Saver Card
Product Code. 31703078
Change view
Click to view available options
Unit Size:
Pack of 100
Product Code. unitSize
31703078 Pack of 100
1 options
Product Code. 31703078
Brand: AHLSTROM 8.570.0000.FN
Lipid Saver™ is an innovative specimen collection card that simplifies the collection, transportation, and long-term preservation of fatty acids from whole blood samples at ambient temperature.
Lipid Saver™ features five printed 12 mm circles, each capable of holding up to 70 μl of blood, allowing for a maximum collection of 350 μl. Compatible with both manual and semi-automated punching systems, Lipid Saver™ seamlessly integrates with current dried blood spot (DBS) technologies. The collection paper is chemically treated with a proprietary stabilizing solution, preventing oxidative degradation of the analytes and ensuring sample integrity over extended periods.
Key Areas Of Use Include:
  • Lipidomics
  • Well-being
  • Diseases Prevention
  • Therapeutic drug monitoring
TRUSTED_SUSTAINABILITY
Specifications
Product Type Specimen Collection and Transport Card
Closure Included Yes
Graduated No
Material Pure Absorbent Fibers
Shape Rectangular
Sterility Non-sterile
Capacity (Metric) 70 μL/Sample Area
Certifications/Compliance ISO 9001