Learn More
Linoleoyl Carnitine, TRC
Specifications
Specifications
| Chemical Name or Material | Linoleoyl Carnitine |
| CAS | 36816-10-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C25 H45 N O4 |
| Quantity | 10 mg |
| Synonym | 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[[(9Z,12Z)-1-oxo-9,12-octadecadien-1-yl]oxy]-, inner salt, (2R)-, 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxo-9,12-octadecadienyl)oxy]-, inner salt, [R-(Z,Z)]-, 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[[(9Z,12Z)-1-oxo-9,12-octadecadienyl]oxy]-, inner salt, (2R)- (9CI), L-Linoleoylcarnitine, Linoleoylcarnitine, Linoleylcarnitine |
| InChI Formula | InChI=1S/C25H45NO4/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-25(29)30-23(21-24(27)28)22-26(2,3)4/h9-10,12-13,23H,5-8,11,14-22H2,1-4H3/b10-9-,13-12-/t23-/m1/s1 |
| SMILES | CCCCC\C=C/C\C=C/CCCCCCCC(=O)O[C@H](CC(=O)[O-])C[N+](C)(C)C |
| IUPAC Name | (3R)-3-[(9Z,12Z)-octadeca-9,12-dienoyl]oxy-4-(trimethylazaniumyl)butanoate |
| Molecular Weight (g/mol) | 423.63 |
| Show More |