Learn More
R&D Systems™ LIH 383
Description
LIH 383 is a potent and highly selective atypical chemokine receptor (ACKR3/CXCR7) agonist. LIH 383 competes directly with CXCL12-AF647 and is more potent in inducing β-arrestin recruitment to ACKR3 (EC50 = 0.61 nM) than the full-length chemokine ligands CXCL12 or CXCL11 (EC50 = 1.2 nM and 2.2 nM, respectively). In a rat ex vivo model, LIH383 increases the availability and signaling of opioid peptides at classical receptors.
Specifications
Specifications
| Product Line | Tocris |
| Purity | 0.95 |
| CAS | 2866266-58-0 |
| Content And Storage | Store at -20°C |
| Molecular Weight (g/mol) | 997.23 |
| Quantity | 1 mg |