missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid Fluorescein Antibody Labeling Kit

Product Code. 18780924 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
30 μg
300 μg
Unit Size:
1 set
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18780924 300 μg 1 set
18035284 30 μg 1 set
18748623 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18780924 Supplier Novus Biologicals™ Supplier No. 3100010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody labeling kit

Fluorescein is a fluorophore widely used in biological analysis, with a pH dependent absorption and emission spectrum. In water it shows an absorption maximum at 494 nm and an emission maximum of 521 nm.

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 300 μg
Conjugate Fluorescein
Kit Contents Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent., 1 vial of Rapid Quencher reagent.
Product Type Rapid Fluorescein Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.