missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Rapid DyLight 633 Antibody Labeling Kit
Shop All Bio Techne ProductsDescription
DyLight 594 generates red fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 593nm, high fluorescence at 618nm (extinction coefficient 8.0 x10 cm-1M-1) and high quantum yield.
- Lightning-Link™ Rapid technology can be used to label small quantities of antibody with 100% recovery
- Easy to use antibody labeling kit with quick labeling times
- From 10μg up to 2mg
- Kit with 30 seconds hands-on time
- No separation steps and no losses
- Scalable technology
- Western Blotting, ELISA, Immunohistochemistry, Immunofluorescence and FACS analysis
Order Info
Quantity
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 300 μg |
| Conjugate | DyLight 633 |
| Product Type | Rapid DyLight 633 Antibody Labeling Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?