missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Rapid DyLight 594 Antibody Labeling Kit
Shop All Bio Techne ProductsDescription
DyLight 594 generates red fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 593nm, high fluorescence at 618nm (extinction coefficient 8.0 x10&superscript_begin;4&superscript_end; cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield.
Order Info
Quantity
Specifications
Specifications
| Product Type | Rapid DyLight 594 Antibody Labeling Kit |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Conjugate | DyLight 594 |
| Quantity | 300 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?