missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid DyLight 594 Antibody Labeling Kit

Product Code. 18706733 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
30 μg
300 μg
Unit Size:
1 set
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18706733 30 μg 1 set
18757303 300 μg 1 set
18063714 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18706733 Supplier Novus Biologicals™ Supplier No. 3240030
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Antibody labeling kit

DyLight 594 generates red fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 593nm, high fluorescence at 618nm (extinction coefficient 8.0 x10&superscript_begin;4&superscript_end; cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield.

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 30 μg
Conjugate DyLight 594
Product Type Rapid DyLight 594 Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.