Learn More
Novus Biologicals™ Lightning-Link (R) Rapid DyLight 488 Antibody Labeling Kit
Product Code. 18729533 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
30 μg
300 μg
Unit Size:
1 set
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18729533 30 μg 1 set
18708853 300 μg 1 set
18700854 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18729533 Supplier Novus Biologicals™ Supplier No. 3220030
  • ONLINE EXCLUSIVE PRICE
    551.00€
    520.80€ / 1 sets
    Save 30.20€
    5% Off
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Applications: Flow Cytometry
DyLight 488 provides green fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 493nm, high fluorescence at 518nm (extinction coefficient 7.0 x10&superscript_begin;4&superscript_end;cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield.
Order Info
Quantity
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 30 μg
Conjugate DyLight 488
Product Type Rapid DyLight 488 Antibody Labeling Kit