missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid DyLight 405 Antibody Labeling Kit

Product Code. 18719943
Change view
Click to view available options
Quantity:
1 mg
300 μg
Unit Size:
1 set
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18719943 1 mg 1mg
18706683 300 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18719943 Supplier Novus Biologicals™ Supplier No. 3210015
  • ONLINE EXCLUSIVE PRICE
    1175.00€
    1110.90€ / 1mg
    Save 64.10€
    5% Off
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibody labeling kit

DyLight 405 provides blue fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 400nm, high fluorescence at 420nm (extinction coefficient 3.0 x10&superscript_begin;4&superscript_end; cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield.

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Product Type Rapid DyLight 405 Antibody Labeling Kit
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Conjugate DyLight 405
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.