Learn More
Novus Biologicals™ Lightning-Link (R) Rapid DyLight 405 Antibody Labeling Kit
Product Code. 18706683 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
300 μg
Unit Size:
1 set
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18706683 300 μg 1 set
18719943 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18706683 Supplier Novus Biologicals™ Supplier No. 3210010
  • ONLINE EXCLUSIVE PRICE
    739.00€
    698.25€ / 1 sets
    Save 40.75€
    5% Off
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Antibody labeling kit
DyLight 405 provides blue fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 400nm, high fluorescence at 420nm (extinction coefficient 3.0 x10&superscript_begin;4&superscript_end; cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield.
Order Info
Quantity
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 300 μg
Conjugate DyLight 405
Product Type Rapid DyLight 405 Antibody Labeling Kit