missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid DyLight 350 Antibody Labeling Kit

Product Code. 18085794 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
30 μg
Unit Size:
1 set
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18085794 1 mg 1mg
18022604 30 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18085794 Supplier Novus Biologicals™ Supplier No. 3200015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody labeling kit

DyLight 350 provides blue fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 353nm, high fluorescence at 432nm (extinction coefficient 1.5 x10&superscript_begin;4&superscript_end;cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield.

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 1 mg
Conjugate DyLight 350
Product Type Rapid DyLight 350 Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.