missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Rapid DyLight 350 Antibody Labeling Kit
Shop All Bio Techne ProductsDescription
DyLight 350 provides blue fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 353nm, high fluorescence at 432nm (extinction coefficient 1.5 x10&superscript_begin;4&superscript_end;cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield.
Order Info
Quantity
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 30 μg |
| Conjugate | DyLight 350 |
| Product Type | Rapid DyLight 350 Antibody Labeling Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?