missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid DyLight 350 Antibody Labeling Kit

Product Code. 18004454 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
300 μg
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18004454 300 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18004454 Supplier Novus Biologicals™ Supplier No. 3200010300UG
  • ONLINE EXCLUSIVE PRICE
    748.00€
    706.65€ / 1 sets
    Save 41.35€
    5% Off
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The easy-to-use, one step procedure allows researchers to covalently label biomolecules with only 30 seconds hands-on time; conjugates are ready to use in less than twenty minutes

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Quantity 300 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.