missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cyanine Dye 5 (Cy5) has a strong absorption peak at 643nm and a secondary absorption peak at 605nm. The maximum emission peak is at 667nm, with an excitation coefficient of 2.5 x10&superscript_begin;5&superscript_end;cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;. Cy5 can be used as an alternative to AlexaFluor 647, and the phycobiliprotein Allophycocyanin.
Order Info
Quantity
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 30 μg |
| Conjugate | Cy5 |
| Product Type | Rapid Cy5 Antibody Labeling Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?