missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Cy5.5 Antibody Labeling Kit
Shop All Bio Techne ProductsDescription
Cyanine Dye 5.5 (Cy5.5) has a strong absorption peak at 675nm. The maximum emission peak is at 694nm, with an extinction coefficient of 2.5 x10&superscript_begin;5&superscript_end;cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;. Cy5.5 can be used as an alternative to Alexa Fluor 680, WellRed DP-3A, DY- 676 and LightCycler Red.
Order Info
Quantity
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 300 μg |
| Conjugate | Cy5.5 |
| Kit Contents | Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent |
| Product Type | Rapid Cy5.5 Antibody Labeling Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?