missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Atto633 Antibody Labeling Kit
Shop All Bio Techne ProductsDescription
Atto633 is one of a new generation of fluorescent labels. It has a strong absorption at 629nm, high fluorescence at 657nm (extinction coefficient 1.3 x10&superscript_begin;5&superscript_end; cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield. Atto633 can be used as a substitute dye for DY-633, LightCycler Red 640, Alexa Fluor 633, and Cy 5.
Order Info
Quantity
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 300 μg |
| Conjugate | Atto 633 |
| Product Type | Rapid Atto633 Antibody Labeling Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?