missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid Atto488 Antibody Labeling Kit

Product Code. 18797233 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
30 μg
300 μg
Unit Size:
1 set
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18797233 30 μg 1 set
18767193 300 μg 1 set
18016164 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18797233 Supplier Novus Biologicals™ Supplier No. 3500030
  • ONLINE EXCLUSIVE PRICE
    567.00€
    535.50€ / 1 sets
    Save 31.50€
    5% Off
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applications: Flow Cytometry

Atto488 is one of a new generation of fluorescent labels, which has been optimised for excitation with an argon laser. It has a strong absorption at 501nm, high fluorescence at 523nm (extinction coefficient 9.0 x10 cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield.

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Product Type Rapid Atto488 Antibody Labeling Kit
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Conjugate Atto 488
Quantity 30 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.