missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid Atto390 Antibody Labeling Kit

Product Code. 18027695 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
30 μg
300 μg
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18027695 30 μg 1 set
18016474 300 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18027695 Supplier Novus Biologicals™ Supplier No. 3490030
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Antibody labeling kit

Atto390 is one of a new generation of fluorescent labels with a coumarin structure. It has a strong absorption at 390nm, high fluorescence at 479nm (extinction coefficient 2.4 x10 cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield.

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 30 μg
Conjugate Atto 390
Product Type Rapid Atto390 Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.